missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase B Antibody (14D7), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59441C
This item is not returnable.
View return policy
Description
Lactate Dehydrogenase B Monoclonal antibody specifically detects Lactate Dehydrogenase B in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Lactate Dehydrogenase B | |
| Monoclonal | |
| DyLight 650 | |
| EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5 | |
| Recombinant human Lactate Dehydrogenase B (1-334aa) purified from E. coli | |
| 0.1 mL | |
| HIF Target Genes, Hypoxia, Mitochondrial Mediated Pathway | |
| 3945 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA, Immunofluorescence | |
| 14D7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction