missing translation for 'onlineSavingsMsg'
Learn More

Lactate Dehydrogenase Antibody, DyLight 550, Novus Biologicals™

Product Code. 18344254 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344254 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344254 Supplier Novus Biologicals Supplier No. NBP306457R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Lactate Dehydrogenase Polyclonal antibody specifically detects Lactate Dehydrogenase in P. falciparum samples. It is validated for ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lactate Dehydrogenase
Applications ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.1, EC 1.1.1.27, lactate dehydrogenase B, LDH heart subunit, LDH-B, LDH-H, L-lactate dehydrogenase B chain, Renal carcinoma antigen NY-REN-46, TRG-5
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Plasmodium falciparum (strain 3D7) Lactate Dehydrogenase (XP_001349989.1; Met1-Ala316).
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline HIF Target Genes, Hypoxia, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 3945
Target Species P. falciparum
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.