missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactate Dehydrogenase A/LDHA Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Lactate Dehydrogenase A/LDHA Polyclonal antibody specifically detects Lactate Dehydrogenase A/LDHA in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Lactate Dehydrogenase A/LDHA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:1000 |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59 |
| Host Species | Rabbit |
| Immunogen | A synthetic phosphorylated peptide around Y10 of human Lactate Dehydrogenase A/LDHA (NP_001128711.1). LIYNL |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?