missing translation for 'onlineSavingsMsg'
Learn More

Lactate Dehydrogenase A/LDHA Mouse anti-Human, DyLight 650, Clone: 1A4, Novus Biologicals™

Product Code. 18407067 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407067 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407067 Supplier Novus Biologicals Supplier No. NBP174023C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Lactate Dehydrogenase A/LDHA Monoclonal antibody specifically detects Lactate Dehydrogenase A/LDHA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lactate Dehydrogenase A/LDHA
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1A4
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Cell proliferation-inducing gene 19 protein, EC 1.1.1, EC 1.1.1.27, GSD11, lactate dehydrogenase A, lactate dehydrogenase M, LDH muscle subunit, LDH1, LDH-A, LDHM, LDH-M, L-lactate dehydrogenase A chain, proliferation-inducing gene 19, Renal carcinoma antigen NY-REN-59
Host Species Mouse
Immunogen Recombinant human Lactate Dehydrogenase A/LDHA (1-332aa) purified from E.coli (NP_005557)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Hypoxia, Lipid and Metabolism, Mitochondrial Mediated Pathway, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3939
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.