missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactase Antibody (CL14164) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26848JF669
This item is not returnable.
View return policy
Description
Lactase Monoclonal antibody specifically detects Lactase in Human samples. It is validated for Immunohistochemistry
Specifications
| Lactase | |
| Monoclonal | |
| Janelia Fluor 669 | |
| EC 3.2.1, LAC, lactase, Lactase-glycosylceramidase, lactase-phlorizin hydrolase, LPH1, LPHlactase-phlorizin hydrolase-1 | |
| This antibody was developed using a synthetic peptide derived from P09848, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cellular Signaling, metabolism | |
| 3938 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry | |
| CL14164 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction