missing translation for 'onlineSavingsMsg'
Learn More

β-Lactamase-IN-1, MedChemExpress

Product Code. p-200101414
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497898

Brand: Medchem Express HY19773

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress β-Lactamase-IN-1 is an inhibitor of β-Lactamase extracted from patent WO2016027249A1, page 77. β-Lactamase-IN-1 can be used to prepare of tricyclic nitrogen containing compound. β-Lactamase-IN-1 can be used for the research of neisseria gonorrhea infection.

β-Lactamase-IN-1 is an inhibitor of β-Lactamase extracted from patent WO2016027249A1, page 77. β-Lactamase-IN-1 can be used to prepare of tricyclic nitrogen containing compound. β-Lactamase-IN-1 can be used for the research of neisseria gonorrhea infection.

Specifications

Chemical Name or Material β-Lactamase-IN-1
CAS 1075237-97-6
Color Khaki
Purity Grade Notes Research
Molecular Formula C11H13N3O4
Quantity 10 mg
Solubility Information DMSO : ≥ 32 mg/mL (127.37 mM)
SMILES O=C1C=NC2=C(N=C(OC)C=C2)N1C(CO)CO
Molecular Weight (g/mol) 251.24
Formula Weight 251.24
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.