missing translation for 'onlineSavingsMsg'
Learn More

β-Lactamase-IN-1, MedChemExpress

Product Code. 16497898
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497898 10 mg 10mg
16463847 50 mg 50mg
16437716 100 mg 100mg
16430368 5 mg 5mg
16401869 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497898 Supplier Medchem Express Supplier No. HY19773
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress β-Lactamase-IN-1 is an inhibitor of β-Lactamase extracted from patent WO2016027249A1, page 77. β-Lactamase-IN-1 can be used to prepare of tricyclic nitrogen containing compound. β-Lactamase-IN-1 can be used for the research of neisseria gonorrhea infection.

β-Lactamase-IN-1 is an inhibitor of β-Lactamase extracted from patent WO2016027249A1, page 77. β-Lactamase-IN-1 can be used to prepare of tricyclic nitrogen containing compound. β-Lactamase-IN-1 can be used for the research of neisseria gonorrhea infection.

Specifications

Chemical Name or Material β-Lactamase-IN-1
CAS 1075237-97-6
Color Khaki
Purity Grade Notes Research
Molecular Formula C11H13N3O4
Quantity 10 mg
Solubility Information DMSO : ≥ 32 mg/mL (127.37 mM)
SMILES O=C1C=NC2=C(N=C(OC)C=C2)N1C(CO)CO
Molecular Weight (g/mol) 251.24
Formula Weight 251.24
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.