missing translation for 'onlineSavingsMsg'
Learn More
Learn More
lactamase, beta 2 Antibody (OTI4F7), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71903AF647
This item is not returnable.
View return policy
Description
lactamase, beta 2 Monoclonal specifically detects lactamase, beta 2 in Human samples. It is validated for Western Blot.
Specifications
| lactamase, beta 2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LACTB2 | |
| Full length recombinant protein of human lactamase, beta 2 (NP_057111) produced in E.coli. | |
| 0.1 mL | |
| 51110.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI4F7 | |
| Western Blot | |
| beta-lactamase-like protein 2, CGI-83, EC 3.-, lactamase, beta 2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction