missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lactalbumin Alpha Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £443.00
Specifications
| Antigen | Lactalbumin Alpha |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18224752
|
Novus Biologicals
NBP2-56836 |
100 μL |
£443.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653718
|
Novus Biologicals
NBP2-56836-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lactalbumin Alpha Polyclonal specifically detects Lactalbumin Alpha in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Lactalbumin Alpha | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 3906 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KCELSQLLKDIDGYGGIALPELICTMFHTSGYDTQAIVENNESTEYGLFQISNKLWCKSS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| alpha-lactalbumin, lactalbumin, alpha-, Lactose synthase B protein, Lysozyme-like protein 7, LYZL7, MGC138521, MGC138523 | |
| LALBA | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title