missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LACRT Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
£155.00 - £386.00
Specifications
| Antigen | LACRT |
|---|---|
| Concentration | 0.54 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17733204
|
Invitrogen™
PA5121805 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17723204
![]() |
Invitrogen™
PA5121804 |
100 μL |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LACRT Polyclonal antibody specifically detects LACRT in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
| LACRT | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q9GZZ8 | |
| 90070 | |
| Glu20-Ala138 (Accession Q9GZZ8), with N-terminal His and GST Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| LACRT |
| 0.54 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Extracellular glycoprotein lacritin; lacritin; LACRT | |
| LACRT | |
| IgG | |
| Protein A, Antigen affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title