Learn More
IKA™ Labworldsoft™ 6 Visc
Product Code. 17217539 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
IKA ROTAVISC Viscometer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17217539 IKA ROTAVISC Viscometer Each
1 options
Product Code. 17217539 Supplier IKA™ Supplier No. 0020101872
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Labworldsoft™ 6 Visc software enables the automation of laboratory processes by connecting ROTAVISC viscometers to other devices, such as a thermostat, through a PC.
  • Enables the automation of laboratory processes by connecting IKA™ ROTAVISC viscometers to other devices, such as a thermostat, through a PC
  • Easily record measurement processes and parameters for analysis and documentation
  • Monitor and control parameters of the sample, such as temperature and pH measurement
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) IKA ROTAVISC Viscometer
For Use With (Application) Data documentation and device control via PC