Learn More
Thermo Scientific™ 2D Labtainer™ BioProcess Container (BPC), ASI™ 28 film, 3 ports, Luer Lock connectors, injection site
Use small volume Labtainers for sampling, harvesting, preparation, storage and transport of process liquids.
Brand: Thermo Scientific™ SS00009-I
Additional Details : Weight : 0.45000kg
Description
This Thermo Scientific™ Labtainer™ BioProcess Container (BPC) 2D system is composed of Thermo Scientific™ ASI™ 28 film, a 4-layer, 12.5 mL, animal origin–free ethyl vinyl acetate film and includes 3 ports (including an injection site). A wide range of capacities, port configurations and tubing styles are available.
- Helps to eliminate post-use cleaning steps required with reusable containers, and to reduce cross-contamination risks
- All Labtainers constructed in an ISO 7 cleanroom environment under cGMP conditions
Applications:
- Harvesting from, feeding into, and collecting samples from bioreactors and fermenters
- Preparing and storing buffer, culture media, and process liquid
- Collecting chromatography feed and fraction
- Harvesting, storing, and transporting bulk drug product and drug precursors
Specifications
Bioprocess Container | |
1 L | |
EVA | |
ASI™ 28 | |
Luer lock connectors, reducer and injection site | |
50/Cs. | |
Irradiated | |
9.8 in. | |
7.8 in. |
Store at room temperature. | |
3 Ports (1 Luer Lock Insert Connection + Cap, 1 Luer Lock Body Connection + Cap, 1 Injection Site) | |
EVA | |
One 4-mil. poly bag | |
Labtainer™ | |
Room Temperature | |
24.9 x 19.8 cm (9.8 x 7.8 in.) | |
24.9 cm | |
19.8 cm |