missing translation for 'onlineSavingsMsg'
Learn More

Dycem™ Laboratory Safety Mats 38 x 45cm

Product Code. 15767079
Encompass_Preferred
Change view
Click to view available options
Color:
Black
Red
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
15767079 Yellow
15747079 Red
15757079 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15767079 Supplier Dycem™ Supplier No. NS02LG8
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anchor items onto any surface with a high quality and long lasting non-slip mat. Dycem™ Laboratory Safety Mats 38 x 45cm offer superb safety and anti-slip properties ideal for use where spillage or damage can occur.

  • Anti-slip properties
  • Ideal for use where spillage or damage can occur
  • Reduce movement and provide bench protection
  • Protect technicians and equipment
  • Available in a range of colors

Specifications

Dimensions (L x W) 38 x 45 cm
Quantity 10/Pk.
Color Yellow
Type Lab Safety Mat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.