missing translation for 'onlineSavingsMsg'
Learn More

Dycem™ Laboratory Safety Mats 14cm Diamter

Product Code. 15737069
Encompass
Click to view available options
Color:
Black
Blue
Red
Silver
Yellow
This item is not returnable. View return policy

Product Code. 15737069

Brand: Dycem™ NS2141

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Anchor items onto any surface with a high quality and long lasting non-slip mat. Dycem™ Laboratory Safety Mats 14cm Diameter offer superb safety and anti-slip properties ideal for use where spillage or damage can occur.

  • Anti-slip properties
  • 14cm diameter
  • Ideal for use where spillage or damage can occur
  • Reduce movement and provide bench protection
  • Protect technicians and equipment
  • Available in a range of colors

Specifications

Dimensions (L x W) 14 x 14 cm
Quantity 50/Pk.
Color Blue
Type Lab Safety Mat
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.