Learn More
Dycem™ Laboratory Safety Jar Openers
Product Code. 15717139
Encompass
Change view
Click to view available options
Color:
Blue
Silver
Yellow
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
15717139 Blue
15727139 Silver
15737139 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15717139 Supplier Dycem™ Supplier No. LS07JO1
This item requires a price update prior to being ordered.
Request a Price for this item here.
Pain free and effortless solution to unscrewing a tight jar. Dycem™ Laboratory Safety Jar Openers aids in opening difficult to remove lids from jars.
  • Provides a firm grip
  • Invaluable daily living aid
  • Works with push down and twist bottles
  • Your grip is maximised thanks to the unique grip bars, and a tight seal when closing is ensured
  • Capable of storing easily in a pocket
  • Can also be used for small cupboard doors and taps
  • Non-toxic
  • Latex free
  • Antimicrobial
  • Simple to use
  • Long lasting
  • Easy to clean
Specifications
Description Circular
Dimensions (L x W) 11.8 x 11.8 cm
Quantity 50/Pk.
Color Blue
Type Lab Safety Bottle Openers