missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dycem™ Laboratory Safety Bottle Openers
Click to view available options
Color:
Blue
Silver
Yellow
Unit Size:
Pack of 50
Description
- Provides a firm grip
- Invaluable daily living aid
- Works with push down and twist bottles
- Your grip is maximised thanks to the unique grip bars, and a tight seal when closing is ensured
- Capable of storing easily in a pocket
- Can also be used for small cupboard doors and taps
- Non-toxic
- Latex free
- Antimicrobial
- Simple to use
- Long lasting
- Easy to clean
Specifications
Specifications
| Description | Circular |
| Dimensions (L x W) | 5.2 x 5.2 cm |
| For Use With (Application) | Opening Bottles |
| Quantity | 50/Pk. |
| Color | Silver |
| Type | Lab Safety Bottle Opener |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction