missing translation for 'onlineSavingsMsg'
Learn More

Dycem™ Laboratory Safety Bottle Openers

Product Code. 15797129
Encompass_Preferred
Change view
Click to view available options
Color:
Blue
Silver
Yellow
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15797129 Silver Pack of 50
15707139 Yellow -
15787129 Blue -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15797129 Supplier Dycem™ Supplier No. NS07BO4

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Effortlessly unscrew tough bottle tops. Dycem™ Laboratory Safety Bottle Openers aids in unscrewing difficult to remove lids and caps from bottles.

  • Provides a firm grip
  • Invaluable daily living aid
  • Works with push down and twist bottles
  • Your grip is maximised thanks to the unique grip bars, and a tight seal when closing is ensured
  • Capable of storing easily in a pocket
  • Can also be used for small cupboard doors and taps
  • Non-toxic
  • Latex free
  • Antimicrobial
  • Simple to use
  • Long lasting
  • Easy to clean

Specifications

Description Circular
Dimensions (L x W) 5.2 x 5.2 cm
For Use With (Application) Opening Bottles
Quantity 50/Pk.
Color Silver
Type Lab Safety Bottle Opener
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.