missing translation for 'onlineSavingsMsg'
Learn More

TerraCycle™ Laboratory PPE and Plastic Recycling Zero Waste Box™
GREENER_CHOICE

Product Code. p-200109928
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
This item is not returnable. View return policy
Do not send in laboratory PPE or plastics which are autoclaved or contaminated with materials considered hazardous.

Product Code. 30282360

Brand: TerraCycle™ CS270LUK

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Do not send in laboratory PPE or plastics which are autoclaved or contaminated with materials considered hazardous.

Recycle Laboratory PPE and plastic using TerraCycle's Zero Waste Box. It includes signage, shipping to and from your location and processing of the materials. Simply fill it up and send it back to TerraCycle when it's full using the pre-paid shipping label affixed to the back of the box.

Use this Zero Waste Box™ to recycle:

  • Disposable PPE (face masks, gloves, garments, beard nets, disposable shoe covers, earplugs, hair nets)
  • Protective eyewear (safety glasses)
  • Rigid lab plastics
  • Flexible lab plastics
  • Disinfectant wipes
  • We accept all types of disposable face masks made from plastic, such as dust masks, FFP2 (or so-called 'N95'), FFP3 and surgical masks.
  • Do not send in:
  • Medical waste. This includes material contaminated with blood, bodily fluids and waste, which should be collected in a hazardous clinical waste bin
  • Lab plastics that are contaminated with hazardous waste
  • Fabric, homemade or paper face masks
  • Wax earplugs
  • Plant-based and biogradable plastics and wipes
  • Autoclaved material
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic)
  • Batteries, pressurised canisters, broken glass

Specifications

Capacity (Metric) 140 L
Capacity (English) 4.9 cu.ft
Color Beige, Green, Red, White
Dimensions (L x W x H) 38 x 38 x 97 cm
Disposable Single-use
Latex Free Yes
Size Large
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.