missing translation for 'onlineSavingsMsg'
Learn More

TerraCycle™ Laboratory PPE and Plastic Recycling Zero Waste Box™
GREENER_CHOICE

Product Code. 30282360
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
30282360 Large Each
30282361 Medium Each
30282358 Small Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Do not send in laboratory PPE or plastics which are autoclaved or contaminated with materials considered hazardous.
Product Code. 30282360 Supplier TerraCycle™ Supplier No. CS270LUK
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Do not send in laboratory PPE or plastics which are autoclaved or contaminated with materials considered hazardous.

Recycle Laboratory PPE and plastic using TerraCycle's Zero Waste Box. It includes signage, shipping to and from your location and processing of the materials. Simply fill it up and send it back to TerraCycle when it's full using the pre-paid shipping label affixed to the back of the box.

Use this Zero Waste Box™ to recycle:

  • Disposable PPE (face masks, gloves, garments, beard nets, disposable shoe covers, earplugs, hair nets)
  • Protective eyewear (safety glasses)
  • Rigid lab plastics
  • Flexible lab plastics
  • Disinfectant wipes
  • We accept all types of disposable face masks made from plastic, such as dust masks, FFP2 (or so-called 'N95'), FFP3 and surgical masks.
  • Do not send in:
  • Medical waste. This includes material contaminated with blood, bodily fluids and waste, which should be collected in a hazardous clinical waste bin
  • Lab plastics that are contaminated with hazardous waste
  • Fabric, homemade or paper face masks
  • Wax earplugs
  • Plant-based and biogradable plastics and wipes
  • Autoclaved material
  • Hazardous waste (sharp, flammable, reactive, corrosive, ignitable, toxic, infectious or pathogenic)
  • Batteries, pressurised canisters, broken glass

Specifications

Capacity (Metric) 140 L
Capacity (English) 4.9 cu.ft
Color Beige, Green, Red, White
Dimensions (L x W x H) 38 x 38 x 97 cm
Disposable Single-use
Latex Free Yes
Size Large
Sterility Non-sterile
Autoclavable Not Autoclavable
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.