missing translation for 'onlineSavingsMsg'
Learn More

Laboratory Buffer Solution, pH 10.01, Reagecon™

Designed for use in laboratory pH measurement. Reagecon™ Laboratory Buffer Solution, pH 10.01 is tested at 20°C to an accuracy level of ±0.02. This calibration and control buffer is specially formulated to ensure its stability and is supplied with a detailed Certificate of Analysis.

Brand:  Reagecon 90507

Code : A0

Additional Details : Weight : 0.26000kg

Product Code. 11716378

  • £25.05 / 250mL
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Reagecon manufactures the most comprehensive range of pH reagents in the world; these are designed to suit all end user requirements. This laboratory grade buffer is manufactured to exacting specifications with an extended shelf life and is supplied with a detailed Certificate of Analysis and comprehensive label.
Specifications

Specifications

0°C
Colorless
Liquid
Characteristic
100°C
10.01 ±0.02
250 mL
Fully miscible in water
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers