missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Laboratory Performance Check Standard For Environmental Analysis, SPEX CertiPrep™
Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
500 mL
Unit Size:
125mL
500mL
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| K | 100μg/mL |
| P | 100μg/mL |
| SiO&subscript_2; | 100μg/mL |
| Al | 20μg/mL |
| As | 20μg/mL |
| B | 20μg/mL |
| Ba | 20μg/mL |
| Be | 20μg/mL |
| Ca | 20μg/mL |
| Cd | 20μg/mL |
| Co | 20μg/mL |
| Cr | 20μg/mL |
| Cu | 20μg/mL |
| Fe | 20μg/mL |
| Li | 20μg/mL |
| Mg | 20μg/mL |
| Mn | 20μg/mL |
| Mo | 20μg/mL |
| Na | 20μg/mL |
| Ni | 20μg/mL |
| Pb | 20μg/mL |
| Sb | 20μg/mL |
| Se | 20μg/mL |
| Sn | 20μg/mL |
| Sr | 20μg/mL |
| Tl | 20μg/mL |
| V | 20μg/mL |
| Zn | 20μg/mL |
| Ag | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Chemical Name or Material | LPC Standard 1 |
| Packaging | Triple-leached LDPE bottle |
| Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
| Technique | AA, ICP |
| For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
| Type | Certified Reference Material |
| Solvent or Matrix | Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride |
| Quantity | 500 mL |
| Grade | Environmental |
| Concentration or Composition (by Analyte or Components) | Various |
Safety and Handling
Product Identifier
- LPC Standard 1
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : nitric acid, hydrofluoric acid
Product intended for laboratory use only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction