missing translation for 'onlineSavingsMsg'
Learn More

Laboratory Performance Check Standard For Environmental Analysis, SPEX CertiPrep™

Product Code. 15618315 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
125 mL
500 mL
Unit Size:
125mL
500mL
This item is not returnable. View return policy

Product Code. 15618315

Brand: SPEX Certiprep LPC1100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
K100μg/mL
P100μg/mL
SiO&subscript_2;100μg/mL
Al20μg/mL
As20μg/mL
B20μg/mL
Ba20μg/mL
Be20μg/mL
Ca20μg/mL
Cd20μg/mL
Co20μg/mL
Cr20μg/mL
Cu20μg/mL
Fe20μg/mL
Li20μg/mL
Mg20μg/mL
Mn20μg/mL
Mo20μg/mL
Na20μg/mL
Ni20μg/mL
Pb20μg/mL
Sb20μg/mL
Se20μg/mL
Sn20μg/mL
Sr20μg/mL
Tl20μg/mL
V20μg/mL
Zn20μg/mL
Ag5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material LPC Standard 1
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix Nitric Acid/Trace Tartaric Acid/Trace Hydrogen Fluoride
Quantity 125 mL
Grade Environmental
Concentration or Composition (by Analyte or Components) Various
compliance-icons
Product Identifier
  • LPC Standard 1
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid, hydrofluoric acid

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.