Learn More
Epredia™ Laboratory Marking Pen, Secureline Marker II
Mark cassettes and microscope slides with the Epredia™ Laboratory Marking Pen.
Brand: Epredia™ 1451
Code : 50
Additional Details : Weight : 0.00900kg Transport : UN number : 1210 Chem class : 3 Pack group : III
Description
- Resistant to common laboratory solvents (water, 10% neutral buffered formalin, alcohol)
- Extra fine point for precise writing
- Non-sterile
- Not made with natural rubber latex
Specifications
Secureline Marker II | |
Black |