Learn More
Cytiva Laboratory Manifold Standard End Cap
Product Code. 17174441
Change view
Click to view available options
:
For use with Laboratory Manifold
Product Code.
17174441 For use with Laboratory Manifold
1 options
Product Code. 17174441 Supplier Cytiva Supplier No. 4899
Only null left
Add to basket
Add to basket
Replacment end cap for microbiology manifold.
  • Lab manifold is designed for flexibility and reduction in cross-contamination
  • Lab manifold features interchangeable adapters for most funnels, and a coupling device to double testing capacity while still enabling sterilization in standard autoclaves
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Laboratory Manifold
Sterility Sterile