Learn More
Juchheim Laborgeraete™ Laboratory Fire Suppression Trough
Product Code. 10163242
Change view
Click to view available options
Color:
Gray
Dimensions (L x W x H):
450 x 450 x 120 mm
Unit Size:
Each
Product Code. Color Dimensions (L x W x H) unitSize
10163242 Gray 450 x 450 x 120 mm Each
1 options
Product Code. 10163242 Supplier Juchheim Laborgeraete™ Supplier No. 00310
Only null left
Add to basket
Add to basket
Stainless steel tray with removable honeycomb insert. For holding up to 12 liters.
  • Extinguishes burning liquids by removing oxygen
  • Stainless steel tray and honeycomb insert
  • With carrying handles
  • Very good to clean
Specifications
Type Storage Tub
Material Stainless Steel
Color Gray
Shape Square
Dimensions (L x W x H) 450 x 450 x 120 mm