missing translation for 'onlineSavingsMsg'
Learn More

FOX2 Antibody, R&D Systems™

Product Code. 18666676 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18666676 100 μg 100µL
18633139 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18666676 Supplier Novus Biologicals Supplier No. NBP305534100ul
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Chicken Polyclonal Antibody

FOX2 Polyclonal specifically detects FOX2 in Human/Mouse/Rat samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOX2
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:2000, Immunocytochemistry/ Immunofluorescence 1:2000
Formulation Supplied as an aliquot of IgY preparation
Gene Alias dJ106I20.3, Fox-1 homolog B, fox-1 homologue, FOX2Fox-2, fxh, hexaribonucleotide binding protein 2, Hexaribonucleotide-binding protein 2, HRNBP2FOX-2, RBM9RTAHNRBP2, Repressor of tamoxifen transcriptional activity, RNA binding motif protein 9, RNA binding protein fox-1 homolog 2, RNA binding protein, fox-1 homolog (C. elegans) 2, RNA-binding motif protein 9, RNA-binding protein 9
Host Species Chicken
Immunogen N-terminal 99 amino acids of human FOX2 expressed in and purified from E. coli
Purification Method IgY purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23543
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Product Type Antibody
Form Purified
Isotype IgY
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.