Learn More
Lenz Laborglasintrumente Laboratory Bottles
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 07105088
Product Code. 10490513
  • £392.70 / Each
Only null left
Estimated Shipment: 20-01-2026
Add to basket
Description
Description
  • Acc. to ISO 4796, with DIN thread
  • Material: DURAN™
  • With easy-to-read scale and large labelling field for easy marking
  • With screw cap and pouring ring (material: PP, blue; temperature resistance: +140 °C)