missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Label Probe Diluent QF

Product Code. 15952334
Change view
Click to view available options
Quantity:
24 mL
Unit Size:
24mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15952334 24 mL 24mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15952334 Supplier Invitrogen™ Supplier No. QVT0513

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Label Probe Diluent QF, 24 mL, is an aqueous solution containing detergent and an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.

Label Probe Diluent QF, 24 mL, is an aqueous solution containing detergent and an individual component of the ViewRNA™ ISH Tissue Assay Kits, 2-Plex, 24 assays.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 24 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.