Learn More
Tenak™ Clip-on Label Holder
Shop All Tenak Products
Click to view available options
Size:
1 Ea.
10/Pk.
20/Pk.
Unit Size:
Each
Pack of 10
Pack of 100
Description
- Can be used for racks for both upright freezers and chest freezers
- Universal and can be used for most commonly sold racks of both stainless steel and aluminum with a material thickness of up to 1.5 mm
- Has a handel lifter which makes it easier to grab the rack handel (works with TENAK racks)
- Labels are not included
- Easy to install - Simply just clip it onto the rack and a barb secures it
Specifications
Specifications
| For Use With (Equipment) | Racks for both upright freezers and chest freezers |
| Size | 20/Pk. |
| Type | Clip-on Label Holder |