missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ UV Light for Purifier™ Delta Biosafety Cabinets

Product Code. 17803880 Shop All Labconco Corporation Products
Change view
Click to view available options
Base Style:
2-pin
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Base Style Unit Size
17803880 2-pin Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17803880 Supplier Labconco™ Supplier No. 1271300
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

UV lamps for Labconco Purifier Delta Biosafety Cabinets.

UV lamp for use with 6' Purifier Delta Biosafety Cabinets.

Specifications

Base Style 2-pin
Gas Fill Argon
Light Source Germicidal UV-C
Lighting Life 6,000 hours
Voltage 100/115/220/230 V
Wattage 30 watts
Wavelength 254 nm
Weight (English) 5 lb.
Weight (Metric) 2.3 Kg
Color Clear
For Use With (Equipment) Purifier Delta Biosafety Cabinet
Length (English) 3 ft.
Material Borosilicate Glass
Product Type UV Bulb
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.