Learn More
Labconco Three Way Adapter
Permits attachment of three ampules to a single valve port.
Brand: Labconco 7762700
Code : 50
Additional Details : Weight : 0.01000kg
Description
Ampules are attached using Ampule Valve Adapters or 1/8 ”surgical tubing (not provided). Cavity in adapter body can be filled with cotton fiber media to help prevent contamination between samples.
Specifications
Adapter |