Learn More
Labconco™ Syringe Kit
Product Code. 15410325 Shop All Labconco Corporation Products
Click to view available options
For Use With (Equipment):
XPert Balance Enclosures
Unit Size:
Each
Product Code. 15410325
Brand: Labconco™ 6924900
For carbon saturation detection
Accessory to FilterMate Portable Exhausters and XPert Balance Systems.
Specifications
For Use With (Equipment) XPert Balance Enclosures
Weight (English) 1 lb.
Weight (Metric) 0.5 Kg
Product Type Syringe Kit
For Use With (Application) Carbon saturation detection, organic vapor, formaldehyde formalin, acid sulfur, ammonia amine