Learn More
Labconco™ Support Shelves: Telescoping Base Stand
For use with Telescoping Base Stands supporting Purifier Logic Biosafety Cabinets and PuriCare Procedure Stations.
POA
Specifications
For Use With (Application) | Provides work surface to support remote equipment |
---|---|
Type | Support Shelf |
Description
For use with Telescoping Base Stands supporting Purifier Logic Biosafety Cabinets and PuriCare Procedure Stations (pre-2014). Support Shelf provides 8.0 in. x 20.0 in. (20 x 51 cm) work surface to support remote equipment such as an aspirator pump within easy access of the biosafety cabinet. Powder-coated steel shelf mounts to left-side tubular frame of the base stand. Includes powder-coated steel shelf and fasteners. Not for use with SoLo Hydraulic Lift Base Stand. Installation is required.Specifications
Provides work surface to support remote equipment | |
Support Shelf |