Learn More
Labconco™ Labconco Soda Acid Trap Insert for Pre-2017 Trap
Product Code. 15103095 Shop All Labconco Corporation Products
Change view
Click to view available options
Acid Trap Insert
Product Code.
15103095 Acid Trap Insert
1 options
Product Code. 15103095 Supplier Labconco™ Supplier No. 7772100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Traps are connected between the evaporator and vacuum pump to provide additional protection to the pump.
Specifications
Product Type Soda Acid Trap Insert
Weight (English) 4.0 lb.
Weight (Metric) 1.8 kg