missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Filters

Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement

Brand:  Labconco™ 3707900

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10109152

  • £695.00 / Each
Estimated Shipment: 27-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement

Replacement HEPA Filter, 99.99% efficient on particles 0.3 micron in size. Not suitable for use with biohazardous materials or volatile organic material. For use with Labconco FilterMate Portable Exhausters, Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, XPert Filtered Balance Stations, XPert and RXPert Filtered Balance Systems.
Specifications

Specifications

99.99%
2, 3, 4 ft. Purifier™ Class I Safety, HEPA Filtered, Vertical Clean Bench and PCR Enclosures
40 lb.
10 to 20 in.
18 in.
10 to 20 x 18 x 18 in. (25.4 to 50.8 x 45.7 x 45.7 cm)
25.4 to 50.8 cm
Filter
HEPA
18 in.
18.1 Kg
3.1 in.
45.7 cm
7.8 cm
45.7 cm
Particulate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers