missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Protector™ Acid and Standard Storage Cabinets Shelf Kits

Product Code. 18764204 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector Acid Storage Cabinet
Width (English) Exterior:
30 in.
48 in.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Width (English) Exterior Unit Size
18764204 Protector Acid Storage Cabinet 30 in. Each
16348705 Protector Acid Storage Cabinet 48 in. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18764204 Supplier Labconco™ Supplier No. 9919002
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Shelf Kits provide additional storage space in Labconco Protector Standard and Acid Base Cabinets.

Interior Shelf Kit provides additional storage space inside Labconco Protector Acid Storage Cabinets. Includes powder-coated steel shelf, acid-resistant PVC tray, and mounting hardware.

Specifications

For Use With (Equipment) Protector Acid Storage Cabinet
Includes Epoxy Coated Steel shelf, Acid-resistant Polyethylene tray and mounting hardware
Width (Metric) Exterior 76 cm
Product Type Shelf Kit
Width (English) Exterior 30 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.