missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ PrimeMate™ Vacuum Pump Oil

Product Code. 15103025 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Hydro-treated Paraffinic Hydrocarbon Oil
Synthetic Hydrocarbon Oil
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material unitSize
15103025 Synthetic Hydrocarbon Oil Each
17104534 Hydro-treated Paraffinic Hydrocarbon Oil Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15103025 Supplier Labconco™ Supplier No. 7541300

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For use with the Labconco PrimeMate Oil Change System

  • Choice of one gallon (3.8L) hydro-treated paraffinic hydrocarbon oil or synthetic hydrocarbon oil
  • Recommended for Labconco general purpose vacuum pumps

Specifications

Product Type Vacuum Pump Oil
Format Oil
For Use With (Equipment) PrimeMate Oil Change System
Material Synthetic Hydrocarbon Oil
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.