Learn More
Labconco™ Manual Valve Gas and Vacuum Tubing Connection Kit for Precise™ Glove Boxes
Product Code. 11711469 Shop All Labconco Corporation Products
Click to view available options
For Use With (Equipment):
Precise Basic Glove Box, Controlled Atmosphere Glove Box
Unit Size:
Each
Product Code. 11711469
Brand: Labconco™ 5245100
Accessory for Precise Glove Boxes
  • Includes flexible vinyl tubing and fittings
  • Connects user-supplied inert gas supply tank and rotary vane vacuum pump to valves located on the glove box
Specifications
For Use With (Equipment) Precise Basic Glove Box, Controlled Atmosphere Glove Box
Weight (English) 20 lb.
Weight (Metric) 9.1 Kg
Product Type Manual Valve Gas and Vacuum Tubing Connection Kit