Learn More
Labconco™ Lids for CentriVap™Cold Traps
Product Code. 16652314 Shop All Labconco Corporation Products
Click to view available options
For Use With (Equipment):
CentriVap Concentrator Cold Trap
Unit Size:
Each
Product Code. 16652314
Brand: Labconco™ 7386700
Only null left
Add to basket
Add to basket
Provides corrosion resistance.
Stainless Steel Lid may be installed in place of the acrylic lid on -50 CentriVap Cold Trap when applications involve solvents which attack acrylic. Included on CentriVap Solvent and Proteomic Systems and -84 and -105 CentriVap Cold Traps.
Specifications
Material Stainless Steel
Type Concentrator Cold Trap Lid
For Use With (Equipment) CentriVap Concentrator Cold Trap