missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Labconco™ Freeze-Drying Ampule Accessories
Shop alle Labconco Corporation produkter
Klik for at se tilgængelige muligheder
Type:
Sealing Torch
Pakningsstørrelse:
Each
Beskrivelse
Freeze drying ampule accessories allow ampules to be used with Labconco FreeZone Freeze Dryers.
- Large rubber split stoppers fit 5, 10, 20, 30, 50, 100 and 125 ml Labconco Serum Bottles (bottles with 20 mm corkage).
- Package of 100.
- Oxygen/Natural Gas Sealing Torch is specifically designed for flame sealing freeze dry ampules.
- Seals all types of heat-resistant glass.
- Connects to natural gas, butane or propane and oxygen with 1/4 in. ID hose connectors.
- Ampule valve adapter provides connection of ampules to drying chambers and manifolds by connecting ampules to valve ports or stainless steel stems.
- Ten per package.
Tekniske data
Tekniske data
| Type | Sealing Torch |
| Material | Stainless Steel |
| For Use With (Application) | Connects to natural gas, butane, or propane and oxygen |
| For Use With (Equipment) | Freeze drying system |
| Weight (English) | 3 lb. |
| Weight (Metric) | 1.4 kg |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion