missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ FilterMate™ Portable Exhauster: Gaseous Contaminants
Removehazardous powders, particulates or vapors from exhaust air stream, then return filtered air to environment by using a Labconco FilterMate™ Portable Exhauster for gaseous contaminants.
£4485.00
Specifications
Product Type | Exhauster |
---|---|
Flow Rate | 250 cfm |
Height (English) | 22.7 in. |
Height (Metric) | 58 cm |
Includes: 8 ft. of 5 in. I.D. gray flexible hose, two clamps and one 5 in. O.D. Upper Transition Adaptor for connection to Demonstration Hood, XPert Balance Enclosure, Protector XVS Ventilation Station or Protector Work Station.
Product Code | Brand | Certifications/Compliance | Hertz | Voltage | Dimensions (L x W x H) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Certifications/Compliance | Hertz | Voltage | Dimensions (L x W x H) | Price | Quantity & Availability | ||
16630463
|
Labconco™
3970021 |
CE | 50 Hz | 230 V | 14 x 23 x 22.7 in. (36 x 58 x 58 cm) |
£4485.00
Each |
Estimated Shipment: 30-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
An accessory for Demonstration Hood, XPert Balance Enclosure, Protector XVS Ventilation Station or Protector Work Station.
- Compact, corrosion-resistant housing of glacier white and light gray, dry powder, epoxy-coated steel with rubber feet
- Uses HEPA filter, carbon-based filter or combination of both filters
- Variable speed, non-sparking motorized impeller, adjustable to optimize airflows
- Negative pressure plenum surrounds positive pressure impeller to capture and refilter unfiltered air
- Front-mounted rocker switch controls power to blower
- Rear-mounted auxiliary electrical outlet for connection to airflow monitor
- Suitable for 2 ft., 3 ft. and 4 ft. wide x 22.75 in. high enclosures
- Comes with IEC 3-wire electrical cord with plug
Models for use with HEPA filter
- Include true bag-in bag-out filter disposal system for safely removing and replacing filter without detaching hose
- Filter efficiency: 250 CFM
Filter efficiency: 250 CFM
Models for use with HEPA and Carbon-based filters
Filter efficiency: 200 CFM
Compliance
UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1
Specifications
Exhauster | |
250 cfm | |
22.7 in. | |
58 cm |