missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Dry Ice Freeze Dry Systems

Product Code. 15163015 Shop All Labconco Corporation Products
Change view
Click to view available options
Capacity (Metric):
1 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15163015 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15163015 Supplier Labconco™ Supplier No. 7522700
  • ONLINE EXCLUSIVE PRICE
    £2730.00
    £2574.18 / Pack of 1
    Save £155.82
    5% Off
 Restricted Item
Estimated Shipment: 31-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for labs with occasional lyophilization needs.

For the laboratory with occasional lyophilization requirements, the Dry Ice Benchtop Freeze Dry System provides a simple, economical means of freeze-drying. Since dry ice cools alcohol or other heat transfer solutions to approximately 75 degrees C or -103 degrees F, the Dry Ice Benchtop Freeze Dry System is ideal for processing materials with low eutectic points. The Dry Ice Benchtop Freeze Dry System is equipped with a center well for dry ice and solvent that serves as a water vapor collector and doubles as a convenient pre-freezing bath. Flasks, serum bottles and ampules may be frozen by dipping and rotating them in the well. Type 304 stainless steel with twelve valves and a single run capacity of 1 liter. Valves accommodate either 1/2 in. or 3/4 in. flask adapters. Includes dry ice/solvent center well with 1.92 liter capacity and cover, 1/2 in. outside diameter port for connection to vacuum pump and 3/8 in. outside diameter port or connection to vacuum gauge.

Requires (not included):

  • Rotary vane direct drive vacuum pump with a minimum displacement of 144 liters/minute, 0.002 mBar ultimate pressure and fitting suitable for 3/4 in. ID vacuum hose
  • Drying accessory: chamber, manifold or tray dryer (some models include a stoppering tray dryer)
  • Freeze dry glassware if not bulk freeze drying
  • Vacuum Gauge
  • Tubing
  • Dry Ice

Specifications

Type Benchtop Freeze Dryer
Capacity (Metric) 1 L
No. of Shelves 0
Temperature Range (Metric) -75°C
Material Stainless Steel
Height (Metric) 24.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.