Learn More
Labconco™ DNA Rotor for CentriVap™ System
Product Code. 15103105 Shop All Labconco Corporation Products
Change view
Click to view available options
Holds:
56 Autosampler Vials
66 Autosampler Vials
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
15103105 56 Autosampler Vials Each
15193095 66 Autosampler Vials Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15103105 Supplier Labconco™ Supplier No. 7824702
Acid-Resistant Thermoplastic Rotors hold tubes at an angle to maximize surface area.
Acid-Resistant Thermoplastic Rotors hold tubes at an angle to maximize surface area. They are easily interchangeable. Sample containers are not included. For use with Labconco CentriVap Vacuum Concentrators.
Specifications
Material Thermoplastic
Type DNA Rotor
For Use With (Equipment) CentriVap™ system
Holds 56 Autosampler Vials
Shipping Weight (English) 3 lb.
Shipping Weight (Metric) 1.4 kg
Weight (English) 3 lb.
Weight (Metric) 1.4 kg