Learn More
Labconco™ Labconco Activated Carbon Solvent Trap Insert for Pre-2017 Trap
Product Code. 15113095 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Activated Carbon Solvent Trap Insert
Unit Size:
Each
Product Code. Product Type Unit Size
15113095 Activated Carbon Solvent Trap Insert Each
1 options
Product Code. 15113095 Supplier Labconco™ Supplier No. 7772600
  • ONLINE EXCLUSIVE PRICE
    £488.00
    £459.94 / Pack of 1
    Save £28.06
    5% Off
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with 7772500 Activated Carbon Solvent Trap.
Traps are connected between the evaporator and vacuum pump to provide additional protection to the pump.
Specifications
Product Type Activated Carbon Solvent Trap Insert
Weight (English) 4.0 lb.
Weight (Metric) 1.8 kg