missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ 6-Place Microtiter Plate Rotor for CentriVap™ Vacuum Concentrator

Product Code. 15103045 Shop All Labconco Corporation Products
Click to view available options
For Use With (Equipment):
CentriVap Vacuum Concentrator
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15103045

Brand: Labconco™ 7553000

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Holds up to 6 standard or 3 deep well microtiter plates

  • Weight: 2.5 lb. (1.1kg)
  • Note: Not for use with CentriVap micro IR Vacuum Concentrator

Specifications

Material Aluminum
Type Microtiter Plate Rotor
For Use With (Equipment) CentriVap Vacuum Concentrator
Holds 6 Standard or 3 Deep-well Microtiter Plate
Weight (English) 2.5 lb.
Weight (Metric) 1.1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.