Learn More
Miele Professional™ Lab Washer
Product Code. 31938165 Shop All Miele Products
Change view
Click to view available options
Model:
PLW 8683 CD AD DC5 CM
PLW 8683 CD ADP DC5 CM
PLW 8683 CD OIL DC5 CM
PLW 8693 DC10
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
31938165 PLW 8683 CD OIL DC5 CM Each
18920720 PLW 8693 DC10 -
31938163 PLW 8683 CD AD DC5 CM Each
31938164 PLW 8683 CD ADP DC5 CM Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 31938165 Supplier Miele Professional™ Supplier No. 12933450
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Offers high-capacity cleaning for up to 128 laboratory bottles per cycle.
  • Flexible and simple modular load carriers and easy load system
  • Maximum user convenience with M Touch Flex full-touch colour display
  • Safety through monitoring-wash pressure and spray arm monitoring
  • Efficient hot air drying-DryPlus
  • Connectivity as standard via WiFi
TRUSTED_SUSTAINABILITY
Specifications
Display Full-color Touch Screen
Material Stainless Steel
Product Type Glassware Washer with Drying System and Steam Condenser
Model PLW 8683 CD OIL DC5 CM
Electrical Requirements 3N AC
Voltage 400 V
Frequency 50 Hz