missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Provide accurate, reproducible results with the Epredia™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-3, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1393-PABX

 View more versions of this product

Product Code. 12619756

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

X-Ray Repair Cross Complementing (XRCC1) plays a role in excision repair of DNA after ionizing irradiation. XRCC1 binds to DNA ligase III through the C-terminal 96 amino acids and to DNA polymerase β through the N-terminal half. In testis XRCC1 is expressed at high levels. Cells with a mutation of this gene exhibit decreased single strand break repair and reduced recombination repair. They show increased double strand breaks, and sister chromatid exchange is increased up to 10-fold. The XRCC1 might serve as a scaffold protein during base excision-repair.

Host Species: Mouse

Clone: 144

Isotype: IgG2a/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human XRCC1 protein

Molecular Weight: 85kDa

Positive Control: Ls174T, or HeLa cells; normal testis

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation (Denatured verified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

XRCC1 (X-Ray Repair Cross Complementing Protein1) Ab-3
Monoclonal
1mg/mL
Mouse
Ls174T, or HeLa cells, normal testis
RUO
Primary
Concentrated
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
144
Unconjugated
Recombinant human XRCC1 protein
200 μL
DNA Repair
Human
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.