Learn More
Epredia™ Lab Vision™ Vimentin Rabbit Monoclonal Antibody
Vimentin, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
Brand: Epredia™ RM-9120-S
Code : LD
Additional Details : Weight : 0.00250kg
Description
Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.Host Species: Rabbit
Clone: SP20
Isotype: IgG
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant protein encoding human vimentin
Molecular Weight: 57-60kDa
Positive Control: Sarcoma and melanoma
Cellular Localization: Cytoplasmic Clone SP20 is excellent for staining of formalin/paraffin with no special pretreatment.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Vimentin | |
Monoclonal | |
Unconjugated | |
Recombinant protein encoding human vimentin | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
SP20 | |
Rabbit | |
1 mL | |
Cytoskeletal Protein | |
Human | |
IgG |