Learn More
Vascular Endothelial Growth Factor (VEGF) Rabbit Polyclonal Antibody, Epredia™
Take advantage of one marker in a comprehensive antibody portfolio that addresses all major disease states with the Epredia™ Vascular Endothelial Growth Factor (VEGF), Rabbit Polyclonal Antibody.
Brand: Epredia RB-9031-RQ
Code : LD
Additional Details : Weight : 0.02000kg
Description
VEGF (vascular endothelial growth factor) is a homodimeric, disulfide-linked glycoprotein involved in angiogenesis which promotes tumor progression and metastasis. This antibody reacts with the 165, 189 and 121 amino acid splice variants of VEGF of human, to a lesser extent, mouse and rat.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: N-terminus
Immunogen: A synthetic peptide derived from N-terminal of human VEGF
Molecular Weight: 19-22kDa (reduced)
Positive Control: Angiosarcoma
Cellular Localization: Cytoplasmic, cell surface and extracellular matrix This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
Vascular Endothelial Growth Factor (VEGF) | |
Polyclonal | |
Rabbit | |
12 mL | |
Angiogenesis |
Immunoprecipitation, Western Blot | |
Unconjugated | |
A synthetic peptide derived from N-terminal of human VEGF | |
IVD | |
Primary |