Learn More
TTF-1 (Thyroid Transcription Factor-1) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ TTF-1 (Thyroid Transcription Factor-1) Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-699-B1
Description
TTF-1(Thyroid transcription factor-1) is a member of the NKx2 family of homeodomain transcription factors. It is expressed in epithelial cells of the thyroid gland and the lung. Nuclei from liver, stomach, pancreas, small intestine, colon, kidney, breast, skin, testes, pituitary, prostate, and adrenal glands are unreactive. TTF-1 is detected in primary lung adenocarcinomas and small cell carcinomas and is absent in colon and breast carcinomas. Staining with TTF-1 antibody is useful for distinguishing between tumors of lung and non-lung origin.Host Species: Mouse
Clone: 8G7G3/1
Isotype: IgG1/κ
Species Reactivity: Human, Mouse and Rat. Shows a broad species reactivity.
Epitope: Not determined
Immunogen: Rat TTF-1 recombinant protein
Molecular Weight: 40kDa
Positive Control: MAD109 cells, normal thyroid or lung
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
Specifications
| TTF-1 (Thyroid Transcription Factor-1) Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 500 μL | |
| Transcription Regulation | |
| Human, Mouse, Rat | |
| IgG2a κ |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 8G7G3/1 | |
| Biotin | |
| Rat TTF-1 recombinant protein | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.